Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclobutane-1,2,3,4-tetracarboxylic dianhydride

Catalog No. 1613305G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-133-05G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-133-05G Supplier Sigma Aldrich Supplier No. 1613305G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cyclobutane-1,2,3,4-tetracarboxylic dianhydride

Specifications

CAS 4415-87-6
Melting Point >300°C (lit.)
Molecular Formula C8H4O6
Linear Formula C8H4O6
MDL Number MFCD00004944
Quantity 250 mg
Synonym 1,2,3,4-Cyclobutanetetracarboxylic dianhydride; Cyclobutanetetracarboxylic acid dianhydride; Cyclobutanetetracarboxylic dianhydride; Maleic anhydride cyclic dimer
Molecular Weight (g/mol) 196.11
Percent Purity ≥94%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.