Learn More
Description
- Spectra of three isomeric trimers of hexafluoropropene have been investigated
- It reacts with primary amines, via indirect substitution of fluorine atoms, to form the corresponding enamines and enimines.hexafluoropropene, trimer may be used in the preparation of photochromic spiroindolinonaphthoxazine derivative
- It may be used in the preparation of 2-trifluoromethyl-3-heptadifluoroisopropyl-2-perfluoropentene-4-(nα,nα-dimethylamino propyl) imino(i)
Specifications
Specifications
| Molecular Formula | C5H7N |
| Linear Formula | C5H7N |
| MDL Number | MFCD00013735 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 81.12 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.