Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclobutanecarbonyl chloride

Catalog No. C9570650G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C9570650G 50 g
C9570610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C9570650G Supplier Sigma Aldrich Supplier No. C9570650G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cyclobutanecarbonyl chloride

Specifications

Boiling Point 60°C (50 mmHg)
CAS 5006-22-4
Quantity 50 g
Molecular Formula C5H7ClO
Linear Formula C4H7COCl
MDL Number MFCD00001319
Synonym Cyclobutancarboxylic acid chloride
Molecular Weight (g/mol) 118.56
Percent Purity 98%
Refractive Index n20/D 1.455 (literature)
Density 1.039 g/mL (at 25°C (literature))
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.