Learn More
Medchemexpress LLC Cyclobutanol | 2919-23-5 | 99.99% | 72.11 | 10 G
SDP
Catalog No. 5000452001
Encompass_Preferred
Cyclobutanol is a biochemical reagent that can be used as a biological material or organic compound for life science-related research.
  • Has synthetic applications of gold-catalyzed ring expansions.
  • Development of novel types of transition metal-catalyzed cascade reactions utilizing small ring systems and their applications.
Catalog No. 50-004-52001 Supplier Medchemexpress LLC Supplier No. HYW01304210G
Cyclobutanol is a biochemical reagent that can be used as a biological material or organic compound for life science-related research.
  • Has synthetic applications of gold-catalyzed ring expansions.
  • Development of novel types of transition metal-catalyzed cascade reactions utilizing small ring systems and their applications.