Learn More
Medchemexpress LLC Cyclobutanol | 2919-23-5 | 99.99% | 72.11 | 25 G
SDP
Catalog No. 5000366490
Encompass_Preferred
Cyclobutanol is a biochemical reagent used in life science research. It has synthetic applications in gold-catalyzed ring expansions and the development of novel transition metal-catalyzed cascade reactions. It is for research use only.
  • Used as a biochemical reagent
  • Suitable for life science related research
  • Appears as a colorless to light yellow liquid
  • Density of 0.921 g/cm³
Catalog No. 50-003-66490 Supplier Medchemexpress LLC Supplier No. 25GHYW013042
Cyclobutanol is a biochemical reagent used in life science research. It has synthetic applications in gold-catalyzed ring expansions and the development of novel transition metal-catalyzed cascade reactions. It is for research use only.
  • Used as a biochemical reagent
  • Suitable for life science related research
  • Appears as a colorless to light yellow liquid
  • Density of 0.921 g/cm³