Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclobutyl bromide, 97%, pure

Catalog No. AC320500050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC320500050 5 g Glass bottle
AC320500010 1 g Glass bottle
AC320500250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC320500050 Supplier Thermo Scientific Chemicals Supplier No. 320500050
Add to Cart

CAS: 4399-47-7 | C4H7Br | 135.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4399-47-7
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.01
MDL Number MFCD00001317
InChI Key KXVUSQIDCZRUKF-UHFFFAOYSA-N
Synonym cyclobutyl bromide, cyclobutane, bromo, cyclobutylbromid, cyclobutylbromide, bromo cyclobutane, bromo-cyclobutane, bromocyclobutane, acmc-209jw1, ksc235o4p, inchi=1/c4h7br/c5-4-2-1-3-4/h4h,1-3h
PubChem CID 78110
IUPAC Name bromocyclobutane
SMILES C1CC(C1)Br

Specifications

Color Colorless
Density 1.4300g/mL
Boiling Point 108.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4776 to 1.4797
Linear Formula C4H7Br
Quantity 5 g
Specific Gravity 1.43
Solubility Information Solubility in water: immiscible
Formula Weight 135.01
Percent Purity 97%
Grade Pure
Physical Form Liquid
Chemical Name or Material Cyclobutyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-530-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.