Learn More
Cayman Chemical Afzelin, Purity: ≥95%, CAS: 482-39-3, 10 mg
SDP
Catalog No. 502989444
Encompass_Preferred
A polyphenolic glycoside flavone with diverse biological activities; scavenges DPPH radicals (IC50 = 6.44 μg/ml); active against S. aureus, P. aeruginosa, S. typhi, C. albicans, C. parapsilosis, and C. neoformans (MICs = 8, 16, 2, 16, 4, and 4 μg/ml, respectively); inhibits the proliferation of A549, SKOV3, and SK-MEL-2 cells (EC50s = 40.6, 34.5, and 33.9 μg/ml, respectively); reduces the number of eosinophils, other inflammatory cells, and total cells, as well as the levels of IL-4, IL-5, and IL-13, in BALF in a mouse model of allergic asthma at 26 mg/kg per day
Catalog No. 50-298-9444 Supplier Cayman Chemical Supplier No. 3113510 MG
A polyphenolic glycoside flavone with diverse biological activities; scavenges DPPH radicals (IC50 = 6.44 μg/ml); active against S. aureus, P. aeruginosa, S. typhi, C. albicans, C. parapsilosis, and C. neoformans (MICs = 8, 16, 2, 16, 4, and 4 μg/ml, respectively); inhibits the proliferation of A549, SKOV3, and SK-MEL-2 cells (EC50s = 40.6, 34.5, and 33.9 μg/ml, respectively); reduces the number of eosinophils, other inflammatory cells, and total cells, as well as the levels of IL-4, IL-5, and IL-13, in BALF in a mouse model of allergic asthma at 26 mg/kg per day