Learn More
Apexbio Technology LLC Cyclocytidine HCl 10212-25-6 200mg
SDP
Supplier:  Apexbio Technology LLC B4869200
Encompass_Preferred
Cyclocytidine HCl (10212-25-6) is a small-molecule nucleoside analog designed to interfere with nucleic acid metabolism and DNA replication Cyclocytidine HCl exerts its biological activity primarily through intracellular conversion into active phosphorylated metabolites that disrupt DNA and RNA synthesis pathways In experimental studies Cyclocytidine HCl demonstrates antitumor activity particularly against leukemia cell lines with reported IC50 values ranging from low micromolar to submicromolar concentrations depending on specific cellular assays and conditions Based on these pharmacological properties Cyclocytidine HCl holds research potential in mechanistic investigations of leukemia pathology and assessment of nucleoside analog efficacy
Catalog No. 50-000-92751