Learn More
Cayman Chemical Cyclocytidine (hydrochloride), Purity: ≥95%, CAS: 10212-25-6, 1 g
SDP
Catalog No. 502941857
Encompass_Preferred
A prodrug form of cytarabine; hydrolyzed to cytarabine in vivo; inhibits the growth of L5178Y leukemia cells in vitro (IC50 = 0.041 µg/ml); inhibits DNA synthesis by inhibiting thymidine incorporation into DNA (IC50 = 110 µg/ml); increases lifespan in an L1210 mouse leukemia model at 3-1,000 mg/kg per day; transiently increases blood pressure in dogs, cats, and rats at 5-100 mg/kg
Catalog No. 50-294-1857 Supplier Cayman Chemical Supplier No. 290141 G
A prodrug form of cytarabine; hydrolyzed to cytarabine in vivo; inhibits the growth of L5178Y leukemia cells in vitro (IC50 = 0.041 µg/ml); inhibits DNA synthesis by inhibiting thymidine incorporation into DNA (IC50 = 110 µg/ml); increases lifespan in an L1210 mouse leukemia model at 3-1,000 mg/kg per day; transiently increases blood pressure in dogs, cats, and rats at 5-100 mg/kg