Learn More
Medchemexpress LLC γ-cyclodextrin | 17465-86-0 | MFCD00009595 | 99.9% | 1297.12 g/mol | C48H80O40 | 1 ML
SDP
Catalog No. 5000299836
Encompass_Preferred
Gamma-cyclodextrin is a cyclic oligosaccharide used as a solubilizing and complexing agent in formulation and biochemical research. It is supplied as high-purity solid or as a 10 mM solution in DMSO for in vitro use.
  • Solubilizes and complexes hydrophobic molecules for assays and formulation.
  • High purity suitable for analytical and preparative workflows.
  • Available as solid or as a ready-to-use 10 mM solution in DMSO.
  • Certificate of analysis and safety data sheet available.
  • Compatible with common in vitro and in vivo co-solvent systems.
Catalog No. 50-002-99836 Supplier Medchemexpress LLC Supplier No. HYW04004010MM/1ML
Gamma-cyclodextrin is a cyclic oligosaccharide used as a solubilizing and complexing agent in formulation and biochemical research. It is supplied as high-purity solid or as a 10 mM solution in DMSO for in vitro use.
  • Solubilizes and complexes hydrophobic molecules for assays and formulation.
  • High purity suitable for analytical and preparative workflows.
  • Available as solid or as a ready-to-use 10 mM solution in DMSO.
  • Certificate of analysis and safety data sheet available.
  • Compatible with common in vitro and in vivo co-solvent systems.