Learn More
Medchemexpress LLC β-cyclodextrin, 6A-(4-methylbenzenesulfonate) | 67217-55-4 | MFCD05864975 | 95.0% | 1289.17 g/mol | C49H76O37S | 1 G
SDP
Catalog No. 5000282615
Encompass_Preferred
Mono-(6-p-toluenesulfonyl)-β-cyclodextrin is a tosylated β-cyclodextrin derivative supplied as a purified powder. It is used as a building block for selective derivatization of cyclodextrins, for host-guest and supramolecular chemistry studies, and as an intermediate in organic synthesis where defined substitution at the 6-position enables controlled functionalization.
  • Tosylated at the 6-position for selective substitution.
  • Useful intermediate for synthesis and derivatization of cyclodextrins.
  • Supports host-guest and supramolecular chemistry studies.
  • Supplied as a powder with specified purity and storage instructions.
  • Compatible with common organic solvents for further reactions.
Catalog No. 50-002-82615 Supplier Medchemexpress LLC Supplier No. HYW1293951G
Mono-(6-p-toluenesulfonyl)-β-cyclodextrin is a tosylated β-cyclodextrin derivative supplied as a purified powder. It is used as a building block for selective derivatization of cyclodextrins, for host-guest and supramolecular chemistry studies, and as an intermediate in organic synthesis where defined substitution at the 6-position enables controlled functionalization.
  • Tosylated at the 6-position for selective substitution.
  • Useful intermediate for synthesis and derivatization of cyclodextrins.
  • Supports host-guest and supramolecular chemistry studies.
  • Supplied as a powder with specified purity and storage instructions.
  • Compatible with common organic solvents for further reactions.