Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-cyclodextrin, 6A-(4-methylbenzenesulfonate) | 67217-55-4 | MFCD05864975 | 95.0% | 1289.17 g/mol | C49H76O37S | 250 MG
SDP

Catalog No. 5000283551
Encompass_Preferred

Mono-(6-p-toluenesulfonyl)-β-cyclodextrin is a monosubstituted β-cyclodextrin derivative with a p-toluenesulfonyl (tosyl) group at the 6-position. It is supplied as a solid for research use and is commonly used as an intermediate for cyclodextrin derivatization and supramolecular chemistry applications.

  • White to off-white solid suitable for laboratory use.
  • Purity 95.0% by 1H NMR.
  • Molecular weight 1289.17 g/mol.
  • Chemical formula C49H76O37S.
  • Optical rotation 111.603° (C=0.01 g/ml, DMF).
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • 1H NMR spectrum consistent with assigned structure.
  • Intended for research use only.

Catalog No. 50-002-83551 Supplier Medchemexpress LLC Supplier No. HYW129395250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mono-(6-p-toluenesulfonyl)-β-cyclodextrin is a monosubstituted β-cyclodextrin derivative with a p-toluenesulfonyl (tosyl) group at the 6-position. It is supplied as a solid for research use and is commonly used as an intermediate for cyclodextrin derivatization and supramolecular chemistry applications.

  • White to off-white solid suitable for laboratory use.
  • Purity 95.0% by 1H NMR.
  • Molecular weight 1289.17 g/mol.
  • Chemical formula C49H76O37S.
  • Optical rotation 111.603° (C=0.01 g/ml, DMF).
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • 1H NMR spectrum consistent with assigned structure.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.