Learn More
Medchemexpress LLC β-cyclodextrin | 7585-39-9 | MFCD00078139 | 100.0% | 1134.98 g/mol | C42H70O35 | 50 G
SDP
Catalog No. 5000240610
Encompass_Preferred
β-Cyclodextrin is a cyclic oligosaccharide composed of seven α-D-glucopyranose units commonly used in research to enhance the solubility and bioavailability of poorly soluble compounds; it has also been reported to exhibit anti-influenza activity. Supplied as a white to off-white solid for laboratory use.
  • Composed of seven glucose units (heptasaccharide).
  • Enhances solubility and bioavailability of hydrophobic agents.
  • High purity (99.95%).
  • Water solubility approximately 7 mg/mL; ultrasonic assistance may be required.
  • Molecular weight 1134.98 g/mol.
  • Powder storage recommended at -20°C (long term) or 4°C (short term).
Catalog No. 50-002-40610 Supplier Medchemexpress LLC Supplier No. HY10720150G
β-Cyclodextrin is a cyclic oligosaccharide composed of seven α-D-glucopyranose units commonly used in research to enhance the solubility and bioavailability of poorly soluble compounds; it has also been reported to exhibit anti-influenza activity. Supplied as a white to off-white solid for laboratory use.
  • Composed of seven glucose units (heptasaccharide).
  • Enhances solubility and bioavailability of hydrophobic agents.
  • High purity (99.95%).
  • Water solubility approximately 7 mg/mL; ultrasonic assistance may be required.
  • Molecular weight 1134.98 g/mol.
  • Powder storage recommended at -20°C (long term) or 4°C (short term).