Learn More
Cayman Chemical β-Cyclodextrin (hydrate), Purity: ≥95%, CAS: 68168-23-0, 100 g
SDP
Catalog No. 502986590
Encompass_Preferred
A cyclic oligosaccharide; has been used to improve the aqueous solubility of various compounds, especially those containing a phenyl group
Catalog No. 50-298-6590 Supplier Cayman Chemical Supplier No. 23387100 G
A cyclic oligosaccharide; has been used to improve the aqueous solubility of various compounds, especially those containing a phenyl group