Learn More
Cayman Chemical Cyclodextrin hydrate 25g
SDP
Catalog No. 502964087
Encompass_Preferred
A cyclic oligosaccharide; has been used to improve the aqueous solubility of various compounds, especially those containing a phenyl group
Catalog No. 50-296-4087 Supplier Cayman Chemical Supplier No. 2338725 G
A cyclic oligosaccharide; has been used to improve the aqueous solubility of various compounds, especially those containing a phenyl group