Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γ- Cyclodextrin, MP Biomedicals™

Catalog No. ICN19005405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19005405 5 g
ICN19005401 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19005405 Supplier MP Biomedicals, Inc Supplier No. 0219005405
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17465-86-0
Molecular Formula C48H80O40
Molecular Weight (g/mol) 1297.13
MDL Number MFCD00009595,MFCD00149574
InChI Key GDSRMADSINPKSL-UHFFFAOYNA-N
Synonym cyclooctaamylose, schardinger gamma-dextrin
PubChem CID 101136809
IUPAC Name 5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

Quantity 5 g
Formula Weight 1297.1
Physical Form Solid
Chemical Name or Material γ- Cyclodextrin
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.