Learn More
MOLECULAR DEPOT LLC Cyclodextrins Kit - 5/2 mL
Catalog No. 5000013451
Encompass_Preferred
CycloScreen Cyclodextrins Kit consists of a kit containing a five 2mL collection of 5 of the most biochemically useful cyclodextrins a-cyclodextrin 2-hydroxypropyl)-a-cyclodextrin methyl-b-cyclodextrin (2-hydroxypropyl)-b-cyclodextrin and(2-hydroxypropyl)-g-cyclodextrin For in vitro research use only
Catalog No. 50-000-13451 Supplier MOLECULAR DEPOT LLC Supplier No. K1010005
CycloScreen Cyclodextrins Kit consists of a kit containing a five 2mL collection of 5 of the most biochemically useful cyclodextrins a-cyclodextrin 2-hydroxypropyl)-a-cyclodextrin methyl-b-cyclodextrin (2-hydroxypropyl)-b-cyclodextrin and(2-hydroxypropyl)-g-cyclodextrin For in vitro research use only