Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclododecanone 99.0+%, TCI America™
SDP

Catalog No. C0462500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0462500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C0462500G Supplier TCI America Supplier No. C0462500G

Chemical Identifiers

CAS 830-13-7
Molecular Formula C12H22O
Molecular Weight (g/mol) 182.307
MDL Number MFCD00003722
InChI Key SXVPOSFURRDKBO-UHFFFAOYSA-N
Synonym cyclododecan-1-one, dsstox_cid_7322, dsstox_rid_78407, dsstox_gsid_27322, cyclododecanon, cdon, cyclododecanone, acmc-209pr1, ksc448s4j
PubChem CID 13246
IUPAC Name cyclododecanone
SMILES C1CCCCCC(=O)CCCCC1

Specifications

Melting Point 62°C
Color White
Boiling Point 113°C
Quantity 500 g
UN Number 3077
Formula Weight 182.31
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclododecanone
EINECSNumber (3)-2379
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.