Learn More
TARGETMOL CHEMICALS INC CYCLODRINE 5MG
SDP
Catalog No. 502581619
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cyclodrine (Cyclopent) is a cholinergic (muscarinic nicotinic) (mAChR and nAChR) receptor antagonist. purity: 98%
Catalog No. 50-258-1619 Supplier TARGETMOL CHEMICALS INC Supplier No. T10914L5MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cyclodrine (Cyclopent) is a cholinergic (muscarinic nicotinic) (mAChR and nAChR) receptor antagonist. purity: 98%