Learn More
Medchemexpress LLC HY-W011100 50mg Medchemexpress, Cyclofenil CAS:2624-43-3 Purity:>98%
SDP
Catalog No. 502083309
Encompass_Preferred
Medchemexpress, HY-W011100 50mg Cyclofenil CAS:2624-43-3 Cyclofenil is a selective estrogen receptor modulator and an ovulation-inducing agent. Cyclofenil shows an inhibitory effect on dengue virus replication in Vero cells with an EC50 of 1.62 μM. Cyclofenil has anti-dengue-virus activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3309 Supplier Medchemexpress LLC Supplier No. HYW01110050MG
Medchemexpress, HY-W011100 50mg Cyclofenil CAS:2624-43-3 Cyclofenil is a selective estrogen receptor modulator and an ovulation-inducing agent. Cyclofenil shows an inhibitory effect on dengue virus replication in Vero cells with an EC50 of 1.62 μM. Cyclofenil has anti-dengue-virus activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.