Learn More
Medchemexpress LLC HY-12784A 5mg , Cycloguanil (hydrochloride) CAS:152-53-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12784A5MG
Encompass_Preferred
HY-12784A 5mg Cycloguanil (hydrochloride) CAS: 152-53-4 Cycloguanil hydrochloride, the active metabolite of Proguanil, acts on malaria schizonts in erythrocytes and hepatocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1346