Learn More
Medchemexpress LLC Cycloguanil hydrochloride | 152-53-4 | 99.9% | 1 ML
SDP
Catalog No. 5000342149
Encompass_Preferred
Cycloguanil hydrochloride is a dihydrofolate reductase (DHFR) inhibitor. It blocks the folate metabolic pathway, affecting nucleotide synthesis and interfering with DNA replication. Cycloguanil inhibits DHFR in *Plasmodium*, making it useful in malaria research. It also potently inhibits DHFR in human cancer cells and blocks the transcriptional activity of STAT3, demonstrating anticancer activity.
  • Inhibits dihydrofolate reductase (DHFR)
  • Blocks folate metabolic pathway
  • Interferes with DNA replication
  • Useful in malaria research
  • Exhibits anticancer activity
  • Potently inhibits transcriptional activity of STAT3
Catalog No. 50-003-42149 Supplier Medchemexpress LLC Supplier No. HY12784A10MM/1ML
Cycloguanil hydrochloride is a dihydrofolate reductase (DHFR) inhibitor. It blocks the folate metabolic pathway, affecting nucleotide synthesis and interfering with DNA replication. Cycloguanil inhibits DHFR in *Plasmodium*, making it useful in malaria research. It also potently inhibits DHFR in human cancer cells and blocks the transcriptional activity of STAT3, demonstrating anticancer activity.
  • Inhibits dihydrofolate reductase (DHFR)
  • Blocks folate metabolic pathway
  • Interferes with DNA replication
  • Useful in malaria research
  • Exhibits anticancer activity
  • Potently inhibits transcriptional activity of STAT3