Learn More
Medchemexpress LLC Cycloguanil hydrochloride | 152-53-4 | 99.9% | 25 MG
SDP
Catalog No. 5000341933
Encompass_Preferred
Cycloguanil hydrochloride is a dihydrofolate reductase (DHFR) inhibitor with an IC50 of 10.8 μM against human DHFR. It functions by blocking the folate metabolic pathway, which affects nucleotide synthesis and interferes with DNA replication. It inhibits DHFR in Plasmodium, making it relevant for malaria research. Additionally, it exhibits anticancer activity by potently inhibiting DHFR in human cancer cells and blocking the transcriptional activity of STAT3.
  • Inhibits dihydrofolate reductase (DHFR)
  • Blocks the folate metabolic pathway
  • Interferes with DNA replication and nucleotide synthesis
  • Relevant for malaria research through Plasmodium DHFR inhibition
  • Exhibits anticancer activity in human cancer cells
  • Blocks transcriptional activity of STAT3
Catalog No. 50-003-41933 Supplier Medchemexpress LLC Supplier No. 25MGHY12784A
Cycloguanil hydrochloride is a dihydrofolate reductase (DHFR) inhibitor with an IC50 of 10.8 μM against human DHFR. It functions by blocking the folate metabolic pathway, which affects nucleotide synthesis and interferes with DNA replication. It inhibits DHFR in Plasmodium, making it relevant for malaria research. Additionally, it exhibits anticancer activity by potently inhibiting DHFR in human cancer cells and blocking the transcriptional activity of STAT3.
  • Inhibits dihydrofolate reductase (DHFR)
  • Blocks the folate metabolic pathway
  • Interferes with DNA replication and nucleotide synthesis
  • Relevant for malaria research through Plasmodium DHFR inhibition
  • Exhibits anticancer activity in human cancer cells
  • Blocks transcriptional activity of STAT3