Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Cyclohexadiene)iron tricarbonyl, 97+%, Thermo Scientific™

Catalog No. AA3940302
Change view
Click to view available options
0.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA3940302 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3940302 Supplier Thermo Scientific Chemicals Supplier No. 3940302

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 12152-72-6
Molecular Formula C9H8FeO3
Quantity 0.5g
MDL Number MFCD00070435
UN Number UN3281
Sensitivity Air Sensitive
Synonym cyclohexadiene iron tricarbonyl, cyclohexadieneiron tricarbonyl, tricarbonylcyclohexadiene iron, cyclohexadiene irontricarbonyl,97+%, cyclohexadiene iron tricarbonyl, 97+%, 1,3-cyclohexadiene tris carbon monoxide iron
InChI Key OVNCEUCQDGRASI-UHFFFAOYSA-N
SMILES [Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].C1CC=CC=C1
IUPAC Name carbon monoxide;cyclohexa-1,3-diene;iron
Molecular Weight (g/mol) 220.01
PubChem CID 10955061
Formula Weight 220.01
Percent Purity ≥97%
Chemical Name or Material (Cyclohexadiene)iron tricarbonyl
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
Precautionary Statement GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, LIQUID, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.