Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclohexane, 1,2,3,4,5,6-hexabromo | 1837-91-8 | MFCD00059127 | 98.0% | 557.54 | C6H6Br6 | 100 MG
SDP

Catalog No. 5000240419
Encompass_Preferred

Benzene hexabromide (cyclohexane, 1,2,3,4,5,6-hexabromo) is a brominated solid used as a biochemical reagent. It acts as a potent inhibitor of JAK2 tyrosine kinase autophosphorylation and is supplied at high purity for laboratory research applications.

  • Chemical formula: C6H6Br6.
  • Molecular weight: 557.54.
  • Purity: 98.0%.
  • Physical form: Solid.
  • CAS number: 1837-91-8.
  • Pack size: 100 MG.
  • Activity: Potent inhibitor of JAK2 tyrosine kinase autophosphorylation.
  • Intended use: For research use only.

Catalog No. 50-002-40419 Supplier Medchemexpress LLC Supplier No. HY107594100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzene hexabromide (cyclohexane, 1,2,3,4,5,6-hexabromo) is a brominated solid used as a biochemical reagent. It acts as a potent inhibitor of JAK2 tyrosine kinase autophosphorylation and is supplied at high purity for laboratory research applications.

  • Chemical formula: C6H6Br6.
  • Molecular weight: 557.54.
  • Purity: 98.0%.
  • Physical form: Solid.
  • CAS number: 1837-91-8.
  • Pack size: 100 MG.
  • Activity: Potent inhibitor of JAK2 tyrosine kinase autophosphorylation.
  • Intended use: For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.