Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclohexane-1,2,3,4,5,6-hexaol | 6917-35-7 | MFCD00077932 | 98.0% | 180.16 g·mol⁻1 | C6H12O6 | 50 G
SDP

Catalog No. 5000301990
Encompass_Preferred

Cyclohexane-1,2,3,4,5,6-hexaol (inositol) is a six-hydroxyl cyclic sugar alcohol used in biochemical research and formulation work. It participates in cell signaling, osmoregulation, and lipid metabolism, and is supplied for laboratory and nutritional applications.

  • Purity 98.0%.
  • Molecular formula C6H12O6.
  • Molecular weight 180.16 g·mol⁻1.
  • White to off-white solid appearance.
  • Stable under recommended storage conditions for powder: -20°C (3 years) or 4°C (2 years).
  • Suitable as a reagent for metabolic and cellular studies.

Catalog No. 50-003-01990 Supplier Medchemexpress LLC Supplier No. HYW02126550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Cyclohexane-1,2,3,4,5,6-hexaol (inositol) is a six-hydroxyl cyclic sugar alcohol used in biochemical research and formulation work. It participates in cell signaling, osmoregulation, and lipid metabolism, and is supplied for laboratory and nutritional applications.

  • Purity 98.0%.
  • Molecular formula C6H12O6.
  • Molecular weight 180.16 g·mol⁻1.
  • White to off-white solid appearance.
  • Stable under recommended storage conditions for powder: -20°C (3 years) or 4°C (2 years).
  • Suitable as a reagent for metabolic and cellular studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.