Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclohexane-1,3-dione | 504-02-9 | 112.13 | 100 G
SDP

Catalog No. 5000460353
Encompass_Preferred

Cyclohexane-1,3-dione is a drug intermediate whose derivatives can exhibit significant tyrosine kinase inhibitory and anti-tumor activities. It is for research use only.

  • Solid appearance
  • White to light yellow color
  • Ships at room temperature in continental US
  • Storage at 4°C, protect from light, stored under nitrogen
  • Soluble in DMSO: ≥ 100 mg/mL

Catalog No. 50-004-60353 Supplier Medchemexpress LLC Supplier No. HYW007466100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Cyclohexane-1,3-dione is a drug intermediate whose derivatives can exhibit significant tyrosine kinase inhibitory and anti-tumor activities. It is for research use only.

  • Solid appearance
  • White to light yellow color
  • Ships at room temperature in continental US
  • Storage at 4°C, protect from light, stored under nitrogen
  • Soluble in DMSO: ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.