Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclohexane, 1,4-bis(bromomethyl)-, trans- | 57702-84-8 | 95.0% | C8H14Br2 | 250 MG
SDP

Catalog No. 5000357645
Encompass_Preferred

Trans-1,4-Di(bromomethyl)cyclohexane is a chemical compound with the formula C8H14Br2. It presents as a white to off-white solid with a purity of 95.0%. This product is intended for research use only.

  • White to off-white solid appearance
  • Purity of 95.0% (NMR)
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-003-57645 Supplier Medchemexpress LLC Supplier No. 250MGHY47683
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Trans-1,4-Di(bromomethyl)cyclohexane is a chemical compound with the formula C8H14Br2. It presents as a white to off-white solid with a purity of 95.0%. This product is intended for research use only.

  • White to off-white solid appearance
  • Purity of 95.0% (NMR)
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.