Learn More
Medchemexpress LLC Cyclohexane-1,4-diamine (1,4-diaminocyclohexane) | 3114-70-3 | 114.19 | 100 G
SDP
Catalog No. 5000448658
Encompass_Preferred
Cyclohexane-1,4-diamine (also known as 1,4-Diaminocyclohexane) is a PROTAC linker used in the synthesis of PROTACs, including TYD-68. This product is intended for research use only.
  • PROTAC linker
  • Suitable for PROTAC synthesis
  • Example application: TYD-68 synthesis
  • For research use
Catalog No. 50-004-48658 Supplier Medchemexpress LLC Supplier No. HYW007732B100G
Cyclohexane-1,4-diamine (also known as 1,4-Diaminocyclohexane) is a PROTAC linker used in the synthesis of PROTACs, including TYD-68. This product is intended for research use only.
  • PROTAC linker
  • Suitable for PROTAC synthesis
  • Example application: TYD-68 synthesis
  • For research use