Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, ≥99.8%, Laboratory Reagent, Solstice

Catalog No. 60047869 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
4 x 2.5 L
4 x 5 L
Packaging:
Aluminum Bottle
Glass Bottle
Metal Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-869 4 x 5 L Aluminum Bottle
60-047-865 1 L Glass Bottle
60-047-866 2.5 L Glass Bottle
60-047-868 4 x 2.5 L Glass Bottle
60-047-870 5 L Aluminum Bottle
60-047-863 10 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-047-869 Supplier Solstice Supplier No. C1003074X5L

missing translation for 'validMainframeMsg'

Laboratory Reagent,≥99.8%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.4°C
Color Colorless
Density 0.779g/mL at 25°C
Quantity 4 x 5 L
Explosion Limit 0.09
Ignition Point 500°F
Odor Characteristic
Packaging Aluminum Bottle
Physical Form Liquid
Vapor Pressure 168.8mmHg (37.7°C), 77mmHg (20°C)
UN Number UN1145
Beilstein 1900225
Boiling Point 81°C (1013 hPa)
Formula Weight 84.16g/mol
Percent Purity ≥99.8%
Chemical Name or Material Cyclohexane
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P331-P501
DOTInformation class 3 - PG 2 - Cyclohexane
RTECSNumber 203-806-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.