Learn More
Sigma Aldrich Fine Chemicals Biosciences Cyclohexane anhydrous 99.5
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 227048100ML
Encompass
Cyclohexane is a cyclic alkane that predominantly exists in chair conformation due to higher stability.
Catalog No. 50-289-8992