Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, Anhydrous Septum-Sealed Bottle, 99.5%, MilliporeSigma™

Catalog No. 50947686 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-686 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-686 Supplier MilliporeSigma Supplier No. CX22846
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Quantity 1 L
Flash Point −18°C
Packaging Glass Bottle (Pre-packed)
Residue after Evaporation 5 ppm max
Assay Percent Range 99.5% min
Vapor Pressure 124 hPa (24°C)
Boiling Point 81°C (1013 hPa)
Solubility Information 0.05 g/L Soluble
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.