Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, CHROMASOLV™, for HPLC, ≥99.7%, Solstice

Catalog No. 6000471 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-004-71 1 L Glass Bottle
60-004-72 2.5 L Glass Bottle
60-004-75 4 x 2.5 L Glass Bottle
60-004-76 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-004-71 Supplier Solstice Supplier No. 348551L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™, for HPLC,≥99.7%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 4°C to 7°C (literature)
Absorbance max. 0.10 at 235 nm, max. 0.20 at 230 nm, max. 0.50 at 220 nm, max. 1.00 at 210 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.01 at 255 nm, max. 0.03 at 250 nm, max. 0.08 at 240 nm
CAS Max % ≤100.0000%
Density 0.779 g/mL at 25 °C(lit.); 0.780 g/mL at 25 °C
Quantity 1 L
Flash Point −18°C
Packaging Glass Bottle
Linear Formula C6H12
UN Number UN1145
Beilstein 1900225
Boiling Point 81°C
Percent Purity ≥99.7%
Grade HPLC
Chemical Name or Material Cyclohexane, CHROMASOLV™, for HPLC, ≥99.7%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Cyclohexane
EINECSNumber 203-806-2
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.