Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, CHROMASOLV™, for pesticide residue analysis, Solstice

Catalog No. 6000480 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-004-80 2.5 L
60-004-79 1 L
60-046-353 4 x 2.5 L
60-004-81 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-004-80 Supplier Solstice Supplier No. 344962.5L
Add to Cart
Add to Cart

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Density 0.779g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.09
Flash Point −18°C (−.4°F)
Ignition Point 500°F
Vapor Pressure 168.8 mmHg (37.7°C), 77 mmHg (20°C)
UN Number UN1145
Beilstein 1900225
Formula Weight 84.16g/mol
Percent Purity ≥99.5% (GC)
Grade CHROMASOLV
Chemical Name or Material Cyclohexane
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P331-P501
RTECSNumber GU6300000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.