Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, CHROMASOLV™ Plus, for HPLC, Solstice
GSA_VA

Catalog No. 6000478 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-004-78 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-004-78 Supplier Solstice Supplier No. 6504551L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™ Plus, for HPLC,≥99.9%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Density 0.779g/mL at 25°C
Quantity 1 L
Explosion Limit 0.09
Flash Point −18°C (−.4°F)
Ignition Point 500°F
Vapor Pressure 77 mmHg (20°C), 168.8 mmHg (37.7°C)
UN Number UN1145
Beilstein 1900225
Formula Weight 84.16g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV™ Plus
Chemical Name or Material Cyclohexane
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P331-P501
RTECSNumber GU6300000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.