Learn More
Sigma Aldrich Fine Chemicals Biosciences Cyclohexane-d12 99.6 atom10G
Catalog No. 502899545
Encompass
Cyclohexane-d12 a deuterated cyclohexane is a standard purity solvent useful for routine NMR (Nuclear Magnetic Resonance) studies. Its infrared (vapor and liquid phase in the range of 376-4000cm-1) and Raman (liquid phase) spectral investigations have been reported. Ring inversion of cyclohexane-d12 has been studied by recording its deuterium NMR spectrum in the temperature range -36 to 115C. It can be prepared by reacting benzene-d6 and deuterium.
Catalog No. 50-289-9545 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 15186610G
Cyclohexane-d12 a deuterated cyclohexane is a standard purity solvent useful for routine NMR (Nuclear Magnetic Resonance) studies. Its infrared (vapor and liquid phase in the range of 376-4000cm-1) and Raman (liquid phase) spectral investigations have been reported. Ring inversion of cyclohexane-d12 has been studied by recording its deuterium NMR spectrum in the temperature range -36 to 115C. It can be prepared by reacting benzene-d6 and deuterium.