Learn More
Cyclohexane, Reagent, ACS, 99%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Cyclohexane, Reagent, ACS is used as a nonpolar solvent, in the calibration of differential scanning calorimetry and other industrial applications including a chemical intermediate. The distinctive detergent-like odor of cyclohexane is reminiscent of cleaning product and is sometimes used in cleaning products as a fragrance enhancer.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 200 L |
| Packaging | Drum |
| Residue after Evaporation | 0.002% |
| Water | 0.02% |
| UN Number | UN1145 |
| Boiling Point | 81°C |
| Percent Purity | 99% |
| Grade | Reagent |
| Chemical Name or Material | Cyclohexane |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.