Learn More
Sigma Aldrich Fine Chemicals Biosciences Cyclohexane suitable for H1L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 6504551L
Encompass
Cyclohexane a cycloalkane is a combustible liquid with petrol like odor. It can be synthesized by hydrogenation of benzene in the presence of nickel or platinum catalysts. Cyclohexane is the precursor for the synthesis of polyamides. Cyclohexane ring forms the structural unit of many naturally occurring products. The conformational changes of cyclohexane have been investigated over the temperature range of 100-1200C. Conversion of cyclohexane to cyclohexanol and cyclohexanone via oxidation in the presence of Cu-nanoclusters anchored on nanocrystalline Cr2O3 has been reported.
Catalog No. 50-289-8990