Learn More
Medchemexpress LLC Cyclohexaneacetamide, N-(4-methoxyphenyl)- | 340221-20-7 | 99.5% | 247.33 | 1 ML
SDP
Catalog No. 5000363159
Encompass_Preferred
sEH inhibitor-7 (compound c-2) is an inhibitor of soluble epoxide hydrolase (sEH), demonstrating IC50s of 0.15 μM (mouse sEH) and 6.2 μM (human sEH). This compound plays a role in the metabolism of endogenous chemical mediators involved in blood pressure regulation and inflammation. It is supplied as an off-white to light yellow solid, available as a 10 mM * 1 mL solution in DMSO.
  • Inhibits soluble epoxide hydrolase (sEH)
  • IC50s of 0.15 μM (mouse sEH) and 6.2 μM (human sEH)
  • Influences metabolism of endogenous chemical mediators
  • Aids in regulation of blood pressure and inflammation
  • Appears as an off-white to light yellow solid
Catalog No. 50-003-63159 Supplier Medchemexpress LLC Supplier No. HY14813610MM/1ML
sEH inhibitor-7 (compound c-2) is an inhibitor of soluble epoxide hydrolase (sEH), demonstrating IC50s of 0.15 μM (mouse sEH) and 6.2 μM (human sEH). This compound plays a role in the metabolism of endogenous chemical mediators involved in blood pressure regulation and inflammation. It is supplied as an off-white to light yellow solid, available as a 10 mM * 1 mL solution in DMSO.
  • Inhibits soluble epoxide hydrolase (sEH)
  • IC50s of 0.15 μM (mouse sEH) and 6.2 μM (human sEH)
  • Influences metabolism of endogenous chemical mediators
  • Aids in regulation of blood pressure and inflammation
  • Appears as an off-white to light yellow solid