Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclohexaneacetic acid | 5292-21-7 | 99.8% | 142.20 g/mol | C8H14O2 | 25 G
SDP

Catalog No. 5000304214
Encompass_Preferred

Cyclohexaneacetic acid (CAS 5292-21-7) is an organic carboxylic acid supplied as a high-purity solid for use as an analytical standard and research reagent. It is suitable for analytical reference, method development, and organic synthesis applications where a well-characterized carboxylic acid is required.

  • High purity (99.78%) analytical-grade material.
  • Molecular weight 142.20 g/mol and formula C8H14O2 for verification and calculations.
  • Available in multiple package sizes, including 25 g, for small- and larger-scale work.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years) to maintain stability.
  • In-solvent stability: -80°C (up to 6 months) or -20°C (up to 1 month) for prepared solutions.

Catalog No. 50-003-04214 Supplier Medchemexpress LLC Supplier No. 25GHYW018653
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclohexaneacetic acid (CAS 5292-21-7) is an organic carboxylic acid supplied as a high-purity solid for use as an analytical standard and research reagent. It is suitable for analytical reference, method development, and organic synthesis applications where a well-characterized carboxylic acid is required.

  • High purity (99.78%) analytical-grade material.
  • Molecular weight 142.20 g/mol and formula C8H14O2 for verification and calculations.
  • Available in multiple package sizes, including 25 g, for small- and larger-scale work.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years) to maintain stability.
  • In-solvent stability: -80°C (up to 6 months) or -20°C (up to 1 month) for prepared solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.