Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclohexaneacetic acid | 5292-21-7 | MFCD00001518 | 99.8% | 142.2 g/mol | C8H14O2 | 10 G
SDP

Catalog No. 5000301025
Encompass_Preferred

Cyclohexaneacetic acid (CAS 5292-21-7) is a small-molecule carboxylic acid provided as a research-grade powder for biochemical and metabolic research. The compound is reported with high purity and recommended storage conditions for long-term stability.

  • High purity (99.8%).
  • Molecular formula C8H14O2 and molecular weight 142.2 g/mol.
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Stable as a powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Intended for research use only (RUO).

Catalog No. 50-003-01025 Supplier Medchemexpress LLC Supplier No. HYW01865310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclohexaneacetic acid (CAS 5292-21-7) is a small-molecule carboxylic acid provided as a research-grade powder for biochemical and metabolic research. The compound is reported with high purity and recommended storage conditions for long-term stability.

  • High purity (99.8%).
  • Molecular formula C8H14O2 and molecular weight 142.2 g/mol.
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Stable as a powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Intended for research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.