Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclohexaneacetic acid | 5292-21-7 | MFCD00001518 | 99.8% | 142.20 g/mol | C8H14O2 | 5 G
SDP

Catalog No. 5000300822
Encompass_Preferred

Cyclohexaneacetic acid is a research-grade small-molecule carboxylic acid (C8H14O2; 142.20 g/mol) supplied as a solid and used as a chemical intermediate and endogenous metabolite reference in biochemical and pharmacological research, providing defined solubility and storage characteristics for analytical and synthetic workflows.

  • High purity: 99.78% (manufacturer reported).
  • Solid, colorless to off-white physical state.
  • Soluble in DMSO at 100 mg/mL; sonication may be required.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Formulation options documented for in vivo use (e.g., DMSO/PEG300/Tween-80/saline).

Catalog No. 50-003-00822 Supplier Medchemexpress LLC Supplier No. HYW0186535G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclohexaneacetic acid is a research-grade small-molecule carboxylic acid (C8H14O2; 142.20 g/mol) supplied as a solid and used as a chemical intermediate and endogenous metabolite reference in biochemical and pharmacological research, providing defined solubility and storage characteristics for analytical and synthetic workflows.

  • High purity: 99.78% (manufacturer reported).
  • Solid, colorless to off-white physical state.
  • Soluble in DMSO at 100 mg/mL; sonication may be required.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Formulation options documented for in vivo use (e.g., DMSO/PEG300/Tween-80/saline).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.