Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclohexanecarbonyl chloride

Catalog No. 156965100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-696-5100G 100 g
15-696-525G 25 g
15-696-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15-696-5100G Supplier Sigma Aldrich Supplier No. 156965100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Electrochemical reduction of cyclohexanecarbonyl chloride at a hanging mercury drop electrode in acetonitrile has been reported.cyclohexanecarbonyl chloride was used in the preparation of five thiourea derivative ligands having anti-bacterial and anti-yeast activity:
  • (n-(diethylcarbamothioyl)cyclohexanecarboxamide
  • N-(di-n-propylcarbamothioyl)cyclohexanecarboxamide
  • Di-n-butylcarbamothioyl)cyclohexanecarboxamide
  • N-(diphenylcarbamothioyl)cyclohexanecarboxamide
  • N-(morpholine-4-carbonothioyl)cyclohexanecarboxamide

Specifications

CAS 2719-27-9
Density 1.096 g/mL (at 25°C (literature))
Boiling Point 184°C (lit.)
Molecular Formula C7H11ClO
Linear Formula C6H11COCl
Refractive Index n20/D 1.469 (literature)
Quantity 100 g
MDL Number MFCD00001456
Synonym Hexahydrobenzoyl chloride
Molecular Weight (g/mol) 146.61
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.