Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclohexanecarbonyl chloride

Catalog No. 1569655G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-696-55G 5 g
15-696-5100G 100 g
15-696-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15-696-55G Supplier Sigma Aldrich Supplier No. 1569655G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Electrochemical reduction of cyclohexanecarbonyl chloride at a hanging mercury drop electrode in acetonitrile has been reported.cyclohexanecarbonyl chloride was used in the preparation of five thiourea derivative ligands having anti-bacterial and anti-yeast activity:
  • (n-(diethylcarbamothioyl)cyclohexanecarboxamide
  • N-(di-n-propylcarbamothioyl)cyclohexanecarboxamide
  • Di-n-butylcarbamothioyl)cyclohexanecarboxamide
  • N-(diphenylcarbamothioyl)cyclohexanecarboxamide
  • N-(morpholine-4-carbonothioyl)cyclohexanecarboxamide

Specifications

CAS 2719-27-9
Density 1.096 g/mL (at 25°C (literature))
Boiling Point 184°C (lit.)
Molecular Formula C7H11ClO
Refractive Index n20/D 1.469 (literature)
Linear Formula C6H11COCl
MDL Number MFCD00001456
Quantity 5 g
Synonym Hexahydrobenzoyl chloride
Molecular Weight (g/mol) 146.61
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.