Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclohexanecarbonyl-L-leucine | 157116-68-2 | MFCD16377985 | 97.0% | 241.33 | C13H23NO3 | 1 G
SDP

Catalog No. 5000210142
Encompass_Preferred

(Cyclohexanecarbonyl)-L-leucine is a cyclohexanecarbonyl-protected derivative of the amino acid L-leucine supplied as a research reagent. It is provided as a white to off-white solid with defined purity and storage recommendations for laboratory use.

  • Cyclohexanecarbonyl-protected L-leucine derivative.
  • White to off-white solid.
  • Molecular formula C13H23NO3; molecular weight 241.33.
  • Purity 97.0% by supplier specification.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Available in small-scale quantities for research use.

Catalog No. 50-002-10142 Supplier Medchemexpress LLC Supplier No. HYW0463521G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Cyclohexanecarbonyl)-L-leucine is a cyclohexanecarbonyl-protected derivative of the amino acid L-leucine supplied as a research reagent. It is provided as a white to off-white solid with defined purity and storage recommendations for laboratory use.

  • Cyclohexanecarbonyl-protected L-leucine derivative.
  • White to off-white solid.
  • Molecular formula C13H23NO3; molecular weight 241.33.
  • Purity 97.0% by supplier specification.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Available in small-scale quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.