Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanecarboxaldehyde, 97%

Catalog No. AC148671000
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148671000 100 mL Glass bottle
AC148670250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC148671000 Supplier Thermo Scientific Chemicals Supplier No. 148671000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2043-61-0 | C7H12O | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2043-61-0
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00001457
InChI Key KVFDZFBHBWTVID-UHFFFAOYSA-N
Synonym cyclohexanecarboxaldehyde, cyclohexanal, formylcyclohexane, cyclohexanealdehyde, cyclohexanaldehyde, cyclohexylcarboxaldehyde, 1-formylcyclohexane, hexahydrobenzaldehyde, cyclohexylformaldehyde, cyclohexane formaldehyde
PubChem CID 16275
IUPAC Name cyclohexanecarbaldehyde
SMILES O=CC1CCCCC1

Specifications

Color Colorless to Pink
Density 0.9260g/mL
Boiling Point 161.0°C to 163.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4490 to 1.4520
Linear Formula C6H11CHO
Quantity 100 mL
Beilstein 07, 19
Specific Gravity 0.926
Formula Weight 112.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyclohexanecarboxaldehyde, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-057-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.