Learn More
Medchemexpress LLC Cyclohexanecarboxylic acid, 1-phenyl-, 2-(4-morpholinyl)ethyl ester, hydrochloride (1:1) | 75136-54-8 | 99.8% | 353.88 | 1 ML
SDP
Catalog No. 5000387211
Encompass_Preferred
PRE-084 hydrochloride is a highly selective σ1 receptor (S1R) agonist, with an IC50 of 44 nM. It exhibits good neuroprotective effects, can improve motor function and motor neuron survival in mice, and can ameliorate myocardial ischemia-reperfusion injury in rats by activating the Akt-eNOS pathway.
  • Highly selective σ1 receptor (S1R) agonist (IC50 of 44 nM).
  • Exhibits neuroprotective effects.
  • Improves motor function and motor neuron survival in mice.
  • Ameliorates myocardial ischemia-reperfusion injury in rats by activating the Akt-eNOS pathway.
Catalog No. 50-003-87211 Supplier Medchemexpress LLC Supplier No. HY18100A10MM/1ML
PRE-084 hydrochloride is a highly selective σ1 receptor (S1R) agonist, with an IC50 of 44 nM. It exhibits good neuroprotective effects, can improve motor function and motor neuron survival in mice, and can ameliorate myocardial ischemia-reperfusion injury in rats by activating the Akt-eNOS pathway.
  • Highly selective σ1 receptor (S1R) agonist (IC50 of 44 nM).
  • Exhibits neuroprotective effects.
  • Improves motor function and motor neuron survival in mice.
  • Ameliorates myocardial ischemia-reperfusion injury in rats by activating the Akt-eNOS pathway.