Learn More
Selleck Chemical LLC Cyclohexanecarboxylic Acid
SDP
Catalog No. 5000003758
Encompass_Preferred
Cyclohexanecarboxylic Acid a component of the growth stimulant naphthenic acid is the starting reagent for the synthesis of polyketide-type antibiotics Phoslactomycins
Catalog No. 50-000-03758 Supplier Selleck Chemical LLC Supplier No. S62795MG
Cyclohexanecarboxylic Acid a component of the growth stimulant naphthenic acid is the starting reagent for the synthesis of polyketide-type antibiotics Phoslactomycins