Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclohexanecarboxylic acid, 3-[[(2E)-3-(3,4-dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-1,4,5-trihydroxy-, (1R,3R,4S,5R)- | 906-33-2 | 354.31 | 50 MG
SDP

Catalog No. 5000416966
Encompass_Preferred

Neochlorogenic acid is a natural polyphenolic compound found in dried fruits and other plants. It inhibits the production of TNF-α and IL-1β, and suppresses iNOS and COX-2 protein expression. It also inhibits phosphorylated NF-κB p65 and p38 MAPK activation. This compound reduces lipopolysaccharide (LPS)-induced NO production in BV2 microglia cells, and inhibits M-CSF/RANKL-induced osteoclast differentiation.

  • Purity: 99.80%
  • Appearance: White to off-white solid
  • Soluble in DMSO and H2O
  • Inhibits TNF-α and IL-1β production
  • Suppresses iNOS and COX-2 protein expression
  • Inhibits phosphorylated NF-κB p65 and p38 MAPK activation

Catalog No. 50-004-16966 Supplier Medchemexpress LLC Supplier No. HYN072250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Neochlorogenic acid is a natural polyphenolic compound found in dried fruits and other plants. It inhibits the production of TNF-α and IL-1β, and suppresses iNOS and COX-2 protein expression. It also inhibits phosphorylated NF-κB p65 and p38 MAPK activation. This compound reduces lipopolysaccharide (LPS)-induced NO production in BV2 microglia cells, and inhibits M-CSF/RANKL-induced osteoclast differentiation.

  • Purity: 99.80%
  • Appearance: White to off-white solid
  • Soluble in DMSO and H2O
  • Inhibits TNF-α and IL-1β production
  • Suppresses iNOS and COX-2 protein expression
  • Inhibits phosphorylated NF-κB p65 and p38 MAPK activation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.