Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclohexanemethanol

Catalog No. C1058055G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1058055G 5 g
C105805100G 100 g
C10580525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C1058055G Supplier Sigma Aldrich Supplier No. C1058055G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Cyclohexanemethanol (cyclohexyl methanol) can be used as a starting material for the synthesis of cyclohexanecarboxaldehyde, cyclohexanecarboxylic acid, cyclohexanone, and 1,4-cyclohexadione by photocatalytic oxidation (pco) using titanium dioxide nanoparticles as a catalyst.reagentplus is a registered trademark of sigma-aldrich co
  • Llc

Specifications

CAS 100-49-2
Density 0.928 g/mL (at 25°C (literature))
Boiling Point 181°C (lit.)
Molecular Formula C7H14O
Refractive Index n20/D 1.465 (literature)
Linear Formula C6H11CH2OH
MDL Number MFCD00001510
Quantity 5 g
Synonym (Hydroxymethyl)cyclohexane; Cyclohexyl methanol; Hexahydrobenzyl alcohol; NSC 5288
Molecular Weight (g/mol) 114.19
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.